accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONH

CAS RN 24706-50-1

IUPAC Name: 3-(phenylamino)cyclohex-2-en-1-one
Synonyms: 3-(phenylamino)cyclohex-2-en-1-one
Molecular Weight (g/mol): 187.24
Molecular Formula: C12H13NO
InChi Key: IVONJXTZXFTWNA-UHFFFAOYSA-N
SMILES: O=C1CCCC(NC2=CC=CC=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)