Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Anilino-2-cyclohexen-1-one, 98%, Thermo Scientific™

Catalog No. AAA1375014
Change view
Click to view available options
Quantity:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1375014 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1375014 Supplier Thermo Scientific Chemicals Supplier No. A1375014

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 24706-50-1
Molecular Formula C12H13NO
Quantity 25g
MDL Number MFCD00082654
Beilstein 2103381
Synonym 3-anilino-2-cyclohexen-1-one, 2-cyclohexen-1-one, 3-phenylamino, 3-phenylamino cyclohex-2-en-1-one, 3-phenylamino cyclohex-2-enone, 3-anilinocyclohex-ene-1-one, 3-phenylamino-2-cyclohexen-1-one, 2-cyclohexen-1-one,3-phenylamino, acmc-1cr8y, 3-anilino-2-cyclohexenone, aronis25001
InChI Key IVONJXTZXFTWNA-UHFFFAOYSA-N
SMILES O=C1CCCC(NC2=CC=CC=C2)=C1
IUPAC Name 3-(phenylamino)cyclohex-2-en-1-one
Molecular Weight (g/mol) 187.24
PubChem CID 98795
Formula Weight 187.24
Percent Purity 98%
Chemical Name or Material 3-Anilino-2-cyclohexen-1-one
Show More Show Less
RTECSNumber GW7600000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.