accessibility menu, dialog, popup

UserName

Viewing profile as user:
SBrFFFSOOO

CAS RN 247129-85-7

IUPAC Name: (2-bromoethyl)diphenylsulfanium trifluoromethanesulfonate
Synonyms: (2-bromoethyl)diphenylsulfanium triflate
Molecular Weight (g/mol): 443.29
Molecular Formula: C15H14BrF3O3S2
InChi Key: SAXNFOVPRAUGNK-UHFFFAOYSA-M
SMILES: [O-]S(=O)(=O)C(F)(F)F.BrCC[S+](C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results