accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHH2N

CAS RN 24716-93-6

IUPAC Name: 2-aminocyclohexane-1-carboxylic acid
Synonyms: 2-aminocyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 143.19
Molecular Formula: C7H13NO2
InChi Key: USQHEVWOPJDAAX-UHFFFAOYNA-N
SMILES: NC1CCCCC1C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)