Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,2S)-2-Aminocyclohexanecarboxylic acid, 97%

Catalog No. AAH52779MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52779MD 250 mg
AAH5277903 1 g
AAH5277906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52779MD Supplier Thermo Scientific Chemicals Supplier No. H52779MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 24716-93-6 | C7H13NO2 | 143.186 g/mol

(1S,2S)-2-Aminocyclohexanecarboxylic acid is used in the preparation of enantiomerically pure beta-amino acids. It is also involved in the synthesis of mono and dihydroxylated derivatives of 2-aminocyclohexanecarboxylic acid by Diels-Alder adduct of ethyl(E)-3-nitroacrylate and furan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1S,2S)-2-Aminocyclohexanecarboxylic acid is used in the preparation of enantiomerically pure beta-amino acids. It is also involved in the synthesis of mono and dihydroxylated derivatives of 2-aminocyclohexanecarboxylic acid by Diels-Alder adduct of ethyl(E)-3-nitroacrylate and furan.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24716-93-6
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
MDL Number MFCD01318270
InChI Key USQHEVWOPJDAAX-WDSKDSINSA-N
PubChem CID 7128323
IUPAC Name (1S,2S)-2-aminocyclohexane-1-carboxylic acid
SMILES C1CCC(C(C1)C(=O)O)N

Specifications

Synonym 1s,2s-2-aminocyclohexanecarboxylic acid, 1s,2s-2-aminocyclohexane-1-carboxylic acid, cyclohexanecarboxylic acid, 2-amino-, 1s,2s, 1s,2s-2-aminocyclohexanecarboxylicacid, 1s,2s-2-amino-cyclohexanecarboxylic acid, 2beta-amino-1alpha-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid,2-amino-, 1s,2s, 24716-92-5 1s,2s-2-aminocyclohexane-1-carboxylic acid
Solubility Information Soluble in water.
Quantity 250 mg
Optical Rotation +77° (c=1 in Water)
Formula Weight 143.19
Percent Purity 97%
Chemical Name or Material (1S,2S)-2-Aminocyclohexanecarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.