accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCl

CAS RN 2473-03-2

IUPAC Name: 1-chloroundecane
Synonyms: 1-chloroundecane
Molecular Weight (g/mol): 190.76
Molecular Formula: C11H23Cl
InChi Key: ZHKKNUKCXPWZOP-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results