Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloroundecane 99.0+%, TCI America™
SDP

Catalog No. C059925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C059925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C059925G Supplier TCI America Supplier No. C059925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2473-03-2
Molecular Formula C11H23Cl
Molecular Weight (g/mol) 190.76
MDL Number MFCD00040870
InChI Key ZHKKNUKCXPWZOP-UHFFFAOYSA-N
Synonym Undecyl Chloride
PubChem CID 17186
IUPAC Name 1-chloroundecane
SMILES CCCCCCCCCCCCl

Specifications

Color Yellow
Boiling Point 242°C
Quantity 25 g
UN Number 3082
Formula Weight 190.76
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloroundecane
EINECSNumber (2)-0066&(2)-0068
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.