accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFFFHOOH

CAS RN 247564-73-4

IUPAC Name: (2,3,5-trifluorophenyl)boronic acid
Synonyms: 2,3,5-trifluorophenylboronic acid
Molecular Weight (g/mol): 175.9
Molecular Formula: C6H4BF3O2
InChi Key: IRMUMGKQAXLGHK-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(F)=CC(F)=C1F
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results