Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-Trifluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. T28745G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T28745G Supplier TCI America Supplier No. T28745G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 247564-73-4
Molecular Formula C6H4BF3O2
Molecular Weight (g/mol) 175.901
MDL Number MFCD01863166
InChI Key IRMUMGKQAXLGHK-UHFFFAOYSA-N
Synonym 2,3,5-Trifluorobenzeneboronic Acid
PubChem CID 4078843
IUPAC Name (2,3,5-trifluorophenyl)boronic acid
SMILES B(C1=CC(=CC(=C1F)F)F)(O)O

Specifications

Melting Point 221°C
Color White-Yellow
Quantity 5 g
Formula Weight 175.90
Physical Form Crystalline Powder
Chemical Name or Material 2,3,5-Trifluorophenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.