accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24821-54-3

CH3OH3C

CAS RN 24821-54-3

IUPAC Name: 1-methoxy-4-methylnaphthalene
Synonyms: 1-methoxy-4-methylnaphthalene
Molecular Weight (g/mol): 172.23
Molecular Formula: C12H12O
InChi Key: DUXRFNPXLWUXMT-UHFFFAOYSA-N
SMILES: COC1=C2C=CC=CC2=C(C)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results