accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNHNS

CAS RN 24854-43-1

IUPAC Name: 4-methyl-4,5-dihydro-1H-1,2,4-triazole-5-thione
Synonyms: 4-methyl-2H-1,2,4-triazole-3-thione
Molecular Weight (g/mol): 115.15
Molecular Formula: C3H5N3S
InChi Key: AGWWTUWTOBEQFE-UHFFFAOYSA-N
SMILES: CN1C=NNC1=S
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)