Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Mercapto-4-methyl-4H-1,2,4-triazole 98.0+%, TCI America™
SDP

Catalog No. M122425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M122425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M122425G Supplier TCI America Supplier No. M122425G
Add to Cart
Add to Cart

Specifications

CAS 24854-43-1
Melting Point 167°C
Color White-Yellow
Molecular Formula C3H5N3S
Quantity 25 g
MDL Number MFCD00010069
Synonym 3-mercapto-4-methyl-4h-1,2,4-triazole, 4-methyl-4h-1,2,4-triazole-3-thiol, 3-mercapto-4-methyl-1,2,4-triazole, 4-methyl-1,2,4-triazole-3-thiol, 3h-1,2,4-triazole-3-thione, 2,4-dihydro-4-methyl, 1,4-dihydro-4-methyl-3h-1,2,4-triazole-3-thione, 4-methyl-1,2,4-triazole-3-thione, 2,4-dimethyl-4-methyl-3h-1,2,4-triazole-3-thione, 4-methyl-4h-1,2,4 triazole-3-thiol, 3h-1,2,4-triazole-3-thione, 1,4-dihydro-4-methyl
InChI Key AGWWTUWTOBEQFE-UHFFFAOYSA-N
SMILES CN1C=NNC1=S
IUPAC Name 4-methyl-1H-1,2,4-triazole-5-thione
Molecular Weight (g/mol) 115.154
PubChem CID 1256162
Formula Weight 115.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Mercapto-4-methyl-4H-1,2,4-triazole
Show More Show Less
RTECSNumber XZ5359126
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.