CAS RN 2489-77-2
IUPAC Name:
1,3,3-trimethylthiourea
Synonyms:
trimethyl thiourea
Molecular Weight (g/mol):
118.2
Molecular Formula:
C4H10N2S
InChi Key:
JAEZSIYNWDWMMN-UHFFFAOYSA-N
SMILES:
CNC(=S)N(C)C
1
–
1
of
1
results
1
Trimethylthiourea 98.0+%, TCI America™
CAS: 2489-77-2 Molecular Formula: C4H10N2S Molecular Weight (g/mol): 118.198 MDL Number: MFCD00059148 InChI Key: JAEZSIYNWDWMMN-UHFFFAOYSA-N PubChem CID: 2779938 IUPAC Name: 1,1,3-trimethylthiourea SMILES: CNC(=S)N(C)C