Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylthiourea 98.0+%, TCI America™
SDP

Catalog No. T0667500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0667500G 500 g
T0667100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0667500G Supplier TCI America Supplier No. T0667500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2489-77-2
Molecular Formula C4H10N2S
Molecular Weight (g/mol) 118.198
MDL Number MFCD00059148
InChI Key JAEZSIYNWDWMMN-UHFFFAOYSA-N
PubChem CID 2779938
IUPAC Name 1,1,3-trimethylthiourea
SMILES CNC(=S)N(C)C

Specifications

Melting Point 84°C
Color White-Yellow
Quantity 500 g
Formula Weight 118.20
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylthiourea
EINECSNumber (2)-1766
RTECSNumber YU4900000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.