accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2492-23-1

IUPAC Name: [(2,4-dichlorophenyl)methyl]triphenylphosphanium chloride
Synonyms: [(2,4-dichlorophenyl)methyl]triphenylphosphanium chloride
Molecular Weight (g/mol): 457.76
Molecular Formula: C25H20Cl3P
InChi Key: FWBSWSPGFNAXPP-UHFFFAOYSA-M
SMILES: [Cl-].ClC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C(Cl)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results