Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2,4-Dichlorobenzyl)triphenylphosphonium Chloride 98.0+%, TCI America™
SDP

Catalog No. D29075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29075G 5 g
D290725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29075G Supplier TCI America Supplier No. D29075G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2492-23-1
Molecular Formula C25H20Cl3P
Molecular Weight (g/mol) 457.759
MDL Number MFCD00031641
InChI Key FWBSWSPGFNAXPP-UHFFFAOYSA-M
Synonym 2,4-dichlorobenzyl triphenylphosphonium chloride, 2,4-dichlorobenzyltriphenylphosphonium chloride, phosphonium, 2,4-dichlorobenzyl triphenyl-, chloride, 2,4-dichlorophenyl methyl triphenylphosphonium chloride, phosphonium, 2,4-dichlorophenyl methyl triphenyl-, chloride, triphenyl 2,4-dichlorobenzyl phosphonium chloride, 2,4-dichlorophenyl methyl triphenylphosphanium chloride, phosphonium,4-dichlorobenzyl triphenyl-, chloride, phosphonium,4-dichlorophenyl methyl triphenyl-, chloride
PubChem CID 196999
IUPAC Name (2,4-dichlorophenyl)methyl-triphenylphosphanium;chloride
SMILES C1=CC=C(C=C1)[P+](CC2=C(C=C(C=C2)Cl)Cl)(C3=CC=CC=C3)C4=CC=CC=C4.[Cl-]

Specifications

Melting Point 249°C
Color White
Quantity 5 g
Formula Weight 457.76
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (2,4-Dichlorobenzyl)triphenylphosphonium Chloride
RTECSNumber TA2200000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.