accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHO

CAS RN 2493-84-7

IUPAC Name: 4-(octyloxy)benzoic acid
Synonyms: 4-(octyloxy)benzoic acid
Molecular Weight (g/mol): 250.34
Molecular Formula: C15H22O3
InChi Key: IALWCYFULVHLEC-UHFFFAOYSA-N
SMILES: CCCCCCCCOC1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)