Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-n-Octyloxybenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. O011725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O011725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O011725G Supplier TCI America Supplier No. O011725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2493-84-7
Molecular Formula C15H22O3
Molecular Weight (g/mol) 250.34
MDL Number MFCD00013993
InChI Key IALWCYFULVHLEC-UHFFFAOYSA-N
Synonym 4-octyloxybenzoic acid, 4-n-octyloxybenzoic acid, 4-octyloxy benzoic acid, p-octyloxybenzoic acid, p-octoxybenzoic acid, p-n-octyloxybenzoic acid, p-octyloxy benzoic acid, benzoic acid, 4-octyloxy, p-n-octyloxy benzoic acid, benzoic acid, p-octyloxy
PubChem CID 17231
IUPAC Name 4-(octyloxy)benzoic acid
SMILES CCCCCCCCOC1=CC=C(C=C1)C(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 250.34
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-n-Octyloxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.