accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CCH3OOBr

CAS RN 24943-60-0

IUPAC Name: [2-(benzoyloxy)ethyl]trimethylazanium bromide
Synonyms: [2-(benzoyloxy)ethyl]trimethylazanium bromide
Molecular Weight (g/mol): 288.19
Molecular Formula: C12H18BrNO2
InChi Key: UOWKZCQHFFILJX-UHFFFAOYSA-M
SMILES: [Br-].C[N+](C)(C)CCOC(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results