Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoylcholine Bromide 98.0+%, TCI America™
SDP

Catalog No. B010625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B010625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B010625G Supplier TCI America Supplier No. B010625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 24943-60-0
Molecular Formula C12H18BrNO2
Molecular Weight (g/mol) 288.19
MDL Number MFCD00059969
InChI Key UOWKZCQHFFILJX-UHFFFAOYSA-M
PubChem CID 22495099
IUPAC Name [2-(benzoyloxy)ethyl]trimethylazanium bromide
SMILES [Br-].C[N+](C)(C)CCOC(=O)C1=CC=CC=C1

Specifications

Melting Point 218°C
Color White
Quantity 25 g
Formula Weight 288.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzoylcholine Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.