accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COBrOOHO

CAS RN 24958-42-7

IUPAC Name: 5-(benzyloxy)-2-bromo-4-methoxybenzoic acid
Synonyms: 5-(benzyloxy)-2-bromo-4-methoxybenzoic acid
Molecular Weight (g/mol): 337.17
Molecular Formula: C15H13BrO4
InChi Key: FBTDEMXQQIWHKX-UHFFFAOYSA-N
SMILES: COC1=C(OCC2=CC=CC=C2)C=C(C(O)=O)C(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results