accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 249609-49-2

IUPAC Name: N1-{[1,1'-biphenyl]-4-yl}-N1-{4-[bis(3,5-dimethylphenyl)amino]phenyl}-N4,N4-bis(3,5-dimethylphenyl)benzene-1,4-diamine
Synonyms: N1-{[1,1'-biphenyl]-4-yl}-N1-{4-[bis(3,5-dimethylphenyl)amino]phenyl}-N4,N4-bis(3,5-dimethylphenyl)benzene-1,4-diamine
Molecular Weight (g/mol): 768.06
Molecular Formula: C56H53N3
InChi Key: XPHQVAAGYDLYOC-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1)N(C1=CC=C(C=C1)N(C1=CC=C(C=C1)N(C1=CC(C)=CC(C)=C1)C1=CC(C)=CC(C)=C1)C1=CC=C(C=C1)C1=CC=CC=C1)C1=CC(C)=CC(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results