accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHN(R)(S)OHO(R)(R)OH(R)OHHOHOO

CAS RN 24967-27-9

IUPAC Name: (2R,3R,4S)-3-acetamido-4-hydroxy-2-[(1R,2R)-1,2,3-trihydroxypropyl]-3,4-dihydro-2H-pyran-6-carboxylic acid
Synonyms: (4S,5R,6R)-5-acetamido-4-hydroxy-6-[(1R,2R)-1,2,3-trihydroxypropyl]-5,6-dihydro-4H-pyran-2-carboxylic acid
Molecular Weight (g/mol): 291.26
Molecular Formula: C11H17NO8
InChi Key: JINJZWSZQKHCIP-UFGQHTETSA-N
SMILES: CC(=O)N[C@@H]1[C@@H](O)C=C(O[C@H]1[C@H](O)[C@H](O)CO)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results