Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-2,3-didehydro-2-deoxyneuraminic Acid, TCI America™
SDP

Catalog No. A22055MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22055MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A22055MG Supplier TCI America Supplier No. A22055MG

Chemical Identifiers

CAS 24967-27-9
Molecular Formula C11H17NO8
Molecular Weight (g/mol) 291.256
MDL Number MFCD00057470
InChI Key JINJZWSZQKHCIP-UFGQHTETSA-N
Synonym 2,3-Dehydro-2-deoxy-N-acetylneuraminic Acid, DANA
PubChem CID 65309
ChEBI CHEBI:28062
IUPAC Name (2R,3R,4S)-3-acetamido-4-hydroxy-2-[(1R,2R)-1,2,3-trihydroxypropyl]-3,4-dihydro-2H-pyran-6-carboxylic acid
SMILES CC(=O)NC1C(C=C(OC1C(C(CO)O)O)C(=O)O)O

Specifications

Melting Point 134°C
Color White-Yellow
Quantity 5 mg
Formula Weight 291.26
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-2,3-didehydro-2-deoxyneuraminic Acid
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.