accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25015-63-8

IUPAC Name: 4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 127.98
Molecular Formula: C6H13BO2
InChi Key: UCFSYHMCKWNKAH-UHFFFAOYSA-N
SMILES: CC1(C)OBOC1(C)C
Boiling Point 
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (7)
  • (5)
Quantity 
  • (2)
  • (1)
  • (4)
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (5)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (7)
  • (5)
  • (2)
  • (1)
  • (4)
  • (1)
  • (4)
  • (2)
  • (5)
  • (5)