Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4,4,5,5-Tetramethyl-1,3,2-Dioxaborolane

Catalog No. 6558565G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-585-65G 5 g
65-585-625G 25 g
65-585-6100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 65-585-65G Supplier Sigma Aldrich Supplier No. 6558565G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4,4,5,5-Tetramethyl-1,3,2-dioxaborolane may be used for:Borylation at the benzylic C-H bond of alkylbenzenes in the presence of a palladium catalyst to form pinacol benzyl boronate.Hydroboration of alkyl or aryl alkynes and alkenes in the presence of transition metal catalysts.Coupling with aryl iodides in the presence of a copper catalyst to form aryl boronates.Asymmetric hydroboration of 1,3-enynes to form chiral allenyl boronates.

Specifications

CAS 25015-63-8
Density 0.882 g/mL at 25°C
Boiling Point 42°C to 43°C (50 mmHg, lit.)
Molecular Formula C6H13BO2
Refractive Index n20/D 1.396
Linear Formula C6H13BO2
MDL Number MFCD00674030
Quantity 5 g
Synonym HBpin; Pinacolborane
Molecular Weight (g/mol) 127.98 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.