accessibility menu, dialog, popup

UserName

Viewing profile as user:
FBr

CAS RN 25017-13-4

IUPAC Name: 1-(2-bromoethyl)-3-fluorobenzene
Synonyms: 1-(2-bromoethyl)-3-fluorobenzene
Molecular Weight (g/mol): 203.05
Molecular Formula: C8H8BrF
InChi Key: GLVSPVSJMYQIPJ-UHFFFAOYSA-N
SMILES: FC1=CC=CC(CCBr)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results