Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3-Fluorophenyl)ethyl Bromide 97.0+%, TCI America™
SDP

Catalog No. B21595G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B21595G Supplier TCI America Supplier No. B21595G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25017-13-4
Molecular Formula C8H8BrF
Molecular Weight (g/mol) 203.054
MDL Number MFCD03095397
InChI Key GLVSPVSJMYQIPJ-UHFFFAOYSA-N
Synonym 1-(2-Bromoethyl)-3-fluorobenzene, 3-Fluorophenethyl Bromide
PubChem CID 573129
IUPAC Name 1-(2-bromoethyl)-3-fluorobenzene
SMILES C1=CC(=CC(=C1)F)CCBr

Specifications

Color Yellow
Boiling Point 94°C
Quantity 5 g
Formula Weight 203.05
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(3-Fluorophenyl)ethyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.