accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNOOF

CAS RN 250371-80-3

IUPAC Name: 1-(2-fluoro-4-nitrophenyl)azepane
Synonyms: 1-(2-fluoro-4-nitrophenyl)azepane
Molecular Weight (g/mol): 238.26
Molecular Formula: C12H15FN2O2
InChi Key: LJVYPBVHNCEVFI-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(F)=C(C=C1)N1CCCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results