accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25038-02-2

IUPAC Name: 2,2,3-trifluoro-3-(trifluoromethyl)oxirane
Synonyms: hexafluoropropylene oxide
Molecular Weight (g/mol): 166.02
Molecular Formula: C3F6O
InChi Key: PGFXOWRDDHCDTE-UHFFFAOYNA-N
SMILES: FC(F)(F)C1(F)OC1(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results