Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(perfluoropropylene Oxide), TCI America™
SDP

Catalog No. P096610G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P096610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P096610G Supplier TCI America Supplier No. P096610G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25038-02-2
Molecular Formula C3F6O
Molecular Weight (g/mol) 166.022
MDL Number MFCD00148042
InChI Key PGFXOWRDDHCDTE-UHFFFAOYSA-N
PubChem CID 9883
IUPAC Name 2,2,3-trifluoro-3-(trifluoromethyl)oxirane
SMILES C1(C(O1)(F)F)(C(F)(F)F)F

Specifications

Color Colorless
Quantity 10 g
Physical Form Liquid
Chemical Name or Material Poly(perfluoropropylene Oxide)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.