CAS RN 25053-15-0
CAS RN 25053-15-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
246.26
Molecular Formula:
C14H14O4
InChi Key:
—
SMILES:
*.C=CCOC(=O)C1=C(C=CC=C1)C(=O)OCC=C
1
–
1
of
1
results
1
Poly(diallyl Phthalate), Average M.W. 65000 (GPC), Thermo Scientific™
CAS: 25053-15-0 Molecular Formula: C14H14O4 Molecular Weight (g/mol): 246.26