accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 250685-44-0

IUPAC Name: methyl 2-{8-[(naphthalen-1-yl)methyl]-4-oxo-1-phenyl-1,3,8-triazaspiro[4.5]decan-3-yl}acetate
Synonyms: methyl 2-[8-(naphthalen-1-ylmethyl)-4-oxo-1-phenyl-1,3,8-triazaspiro[4.5]decan-3-yl]acetate
Molecular Weight (g/mol): 443.55
Molecular Formula: C27H29N3O3
InChi Key: AQMPIDSGLFVVPL-UHFFFAOYSA-N
SMILES: COC(=O)CN1CN(C2=CC=CC=C2)C2(CCN(CC3=C4C=CC=CC4=CC=C3)CC2)C1=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results