Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NNC 63-0532, Tocris Bioscience™
SDP

Catalog No. 178050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-805-0 50 mg
17-801-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-805-0 Supplier R&D Systems Supplier No. 1780/50

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent non-peptide NOP agonist; brain penetrant

Potent non-peptide agonist for the NOP receptor (EC50 = 305 nM, Ki = 7.3 nM); ∽ 12-fold selective over other receptors. Centrally active following systemic administration in vivo.

Chemical Identifiers

CAS 250685-44-0
Molecular Formula C27H29N3O3
Synonym 8-(1-Naphthalenylmethyl)-4-oxo-1-phenyl-1,3,8-triazaspiro[4.5]decane-3-acetic acid, methyl ester

Specifications

Quantity 50 mg
Formula Weight 443.54
Percent Purity >98%
Chemical Name or Material NNC 63-0532
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.