accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHCH3

CAS RN 25078-04-0

IUPAC Name: 2,4-dimethyl-N-phenylaniline
Synonyms: 2,4-dimethyl-N-phenylaniline
Molecular Weight (g/mol): 197.28
Molecular Formula: C14H15N
InChi Key: BWYYRZBQXLCZJL-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(NC2=CC=CC=C2)C=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results