Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethyldiphenylamine 98.0+%, TCI America™
SDP

Catalog No. D339825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D339825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D339825G Supplier TCI America Supplier No. D339825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory reagent with a purity gt;98.0% (GC).

Chemical Identifiers

CAS 25078-04-0
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.281
InChI Key BWYYRZBQXLCZJL-UHFFFAOYSA-N
Synonym N-Phenyl-2,4-xylidine, Phenyl(2,4-xylyl)amine
PubChem CID 10559845
IUPAC Name 2,4-dimethyl-N-phenylaniline
SMILES CC1=CC(=C(C=C1)NC2=CC=CC=C2)C

Specifications

Melting Point 46°C
Color Yellow
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Boiling Point 170°C
Formula Weight 197.28
Percent Purity ≥98.0% (GC)
Grade Not Available
Chemical Name or Material 2,4-Dimethyldiphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.