accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOSH

CAS RN 25103-09-7

IUPAC Name: 6-methylheptyl 2-sulfanylacetate
Synonyms: isooctyl thioglycolate
Molecular Weight (g/mol): 204.33
Molecular Formula: C10H20O2S
InChi Key: RZBBHEJLECUBJT-UHFFFAOYSA-N
SMILES: CC(C)CCCCCOC(=O)CS

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results