Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isooctyl Thioglycolate, MP Biomedicals

Catalog No. ICN202234 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN202234 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN202234 Supplier MP Biomedicals, Inc Supplier No. 0520223405

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25103-09-7
Molecular Formula C10H20O2S
Molecular Weight (g/mol) 204.328
InChI Key RZBBHEJLECUBJT-UHFFFAOYSA-N
Synonym isooctyl thioglycolate, isooctyl mercaptoacetate, acetic acid, mercapto-, isooctyl ester, unii-3404m9zdtb, acetic acid, 2-mercapto-, isooctyl ester, iso-octyl thioglycolate, acmc-20akp0, dsstox_cid_7841, isooctyl 2-mercaptoacetate, dsstox_rid_78587
PubChem CID 34306
IUPAC Name 6-methylheptyl 2-sulfanylacetate
SMILES CC(C)CCCCCOC(=O)CS

Specifications

Quantity 100 g
Formula Weight 204.33
Chemical Name or Material Isooctyl Thioglycolate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.