CAS RN 25109-26-6
IUPAC Name:
selenophene-2-carbaldehyde
Synonyms:
selenophene-2-carbaldehyde
Molecular Weight (g/mol):
159.06
Molecular Formula:
C5H4OSe
InChi Key:
AGBORNIRMFZCGC-UHFFFAOYSA-N
SMILES:
O=CC1=CC=C[Se]1
1
–
1
of
1
results
1
2-Formylselenophene (stabilized with HQ) 98.0+%, TCI America™
CAS: 25109-26-6 Molecular Formula: C5H4OSe Molecular Weight (g/mol): 159.057 InChI Key: AGBORNIRMFZCGC-UHFFFAOYSA-N Synonym: 2-Selenophenecarboxaldehyde PubChem CID: 141174 IUPAC Name: selenophene-2-carbaldehyde SMILES: C1=C[Se]C(=C1)C=O