Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Formylselenophene (stabilized with HQ) 98.0+%, TCI America™
SDP

Catalog No. F07825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07825G 5 g
F07821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07825G Supplier TCI America Supplier No. F07825G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25109-26-6
Molecular Formula C5H4OSe
Molecular Weight (g/mol) 159.057
InChI Key AGBORNIRMFZCGC-UHFFFAOYSA-N
Synonym 2-Selenophenecarboxaldehyde
PubChem CID 141174
IUPAC Name selenophene-2-carbaldehyde
SMILES C1=C[Se]C(=C1)C=O

Specifications

Color Red-Yellow
Boiling Point 85°C
Quantity 5 g
UN Number 3440
Formula Weight 159.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Formylselenophene (stabilized with HQ)
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.