accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2512-56-3

IUPAC Name: 4-octylphenyl 2-hydroxybenzoate
Synonyms: 4-octylphenyl 2-hydroxybenzoate
Molecular Weight (g/mol): 326.44
Molecular Formula: C21H26O3
InChi Key: VNFXPOAMRORRJJ-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=CC=C(OC(=O)C2=CC=CC=C2O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results