Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Octylphenyl Salicylate 95.0+%, TCI America™
SDP

Catalog No. S001625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S001625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001625G Supplier TCI America Supplier No. S001625G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2512-56-3
Molecular Formula C21H26O3
Molecular Weight (g/mol) 326.44
MDL Number MFCD00045760
InChI Key VNFXPOAMRORRJJ-UHFFFAOYSA-N
Synonym Salicylic Acid 4-Octylphenyl Ester
PubChem CID 520185
IUPAC Name 4-octylphenyl 2-hydroxybenzoate
SMILES CCCCCCCCC1=CC=C(OC(=O)C2=CC=CC=C2O)C=C1

Specifications

Melting Point 75°C
Color White
Quantity 25 g
Formula Weight 326.44
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Octylphenyl Salicylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.