CAS RN 251310-57-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Thyroid Hormone Receptor Antagonist, 1-850
The Thyroid Hormone Receptor Antagonist, 1-850, also referenced under CAS 251310-57-3, controls the biological activity of Thyroid Hormone Receptor. This small molecule/inhibitor is primarily used for Neuroscience applications.