accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3P

CAS RN 251320-86-2

IUPAC Name: dicyclohexyl({2'-methyl-[1,1'-biphenyl]-2-yl})phosphane
Synonyms: dicyclohexyl({2'-methyl-[1,1'-biphenyl]-2-yl})phosphane
Molecular Weight (g/mol): 364.51
Molecular Formula: C25H33P
InChi Key: GPVWUKXZFDHGMZ-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1C1=CC=CC=C1P(C1CCCCC1)C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results