Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(-)-3-Amino-1-Boc-Piperidine

Catalog No. 078581GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-858-1GF 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 07-858-1GF Supplier Sigma Aldrich Supplier No. 078581GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(R)-3-Amino-1-Boc-piperidine can be used as a precursor for the preparation of dipeptidyl peptidase IV inhibitors like linagliptin, alogliptin, and other antidiabetic agents.(R)-3-Amino-1-Boc-piperidine can be used to prepare a benzoxazepine derivative named (R)-7-(3,5-dimethoxyphenyl)-N-(piperidin-3-yl)-4-propionyl-2,3,4,5-tetrahydro-1,4-benzoxazepine-9-carboxamide, a potent CBP/P300 bromodomain inhibitor.

Specifications

CAS 251320-86-2
Melting Point 105°C to 109°C
Molecular Formula C25H33P
Linear Formula C25H33P
MDL Number MFCD03094577
Quantity 1 g
Synonym 2-Dicyclohexylphosphino-2'-methylbiphenyl; 2-Methyl-2'-dicyclohexylphosphinobiphenyl; MebiphPCy2
Molecular Weight (g/mol) 364.5 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.