accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOCH3O

CAS RN 25134-21-8

IUPAC Name: 1-methyl-4-oxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
Synonyms: 1-methyl-4-oxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: KNRCVAANTQNTPT-UHFFFAOYNA-N
SMILES: CC12CC(C=C1)C1C2C(=O)OC1=O
Molecular Weight (g/mol) 
  • (4)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (2)
  • (5)
  • (4)
  • (6)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)