Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Nadic Anhydride, MP Biomedicals

Catalog No. ICN216015 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN216015 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN216015 Supplier MP Biomedicals, Inc Supplier No. 0521601580

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25134-21-8
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
InChI Key KNRCVAANTQNTPT-UHFFFAOYNA-N

Specifications

Quantity 100 g
Formula Weight 178.19
Chemical Name or Material Methyl Nadic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.