accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25140-86-7

IUPAC Name: 2-(2-chlorophenoxy)propanoic acid
Synonyms: 2-(2-chlorophenoxy)propanoic acid
Molecular Weight (g/mol): 200.62
Molecular Formula: C9H9ClO3
InChi Key: ZGWNXHRVUJVMCP-UHFFFAOYNA-N
SMILES: CC(OC1=CC=CC=C1Cl)C(O)=O
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)