Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Chlorophenoxy)propionic acid, 98%, Thermo Scientific™

Catalog No. AC209660050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC209660050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC209660050 Supplier Thermo Scientific Chemicals Supplier No. 209660050

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 25140-86-7
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.618
MDL Number MFCD00002644
InChI Key ZGWNXHRVUJVMCP-UHFFFAOYSA-N
Synonym 2-2-chlorophenoxy propanoic acid, 2-2-chlorophenoxy propionic acid, propanoic acid, 2-2-chlorophenoxy, 2-o-chlorophenoxy propionic acid, propionic acid, 2-o-chlorophenoxy, acide o-chlorophenoxy-2 propionique, acide o-chlorophenoxy-2 propionique french, 2-2-chlorophenoxy propionicacid, +/--2-2-chlorophenoxy propionic acid, acmc-1ch4k
PubChem CID 72862
IUPAC Name 2-(2-chlorophenoxy)propanoic acid
SMILES CC(C(=O)O)OC1=CC=CC=C1Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 110°C to 116°C
Color Cream to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula CH3CH(OC6H4Cl)CO2H
Quantity 5 g
Beilstein 06, III, 678
Formula Weight 200.62
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Chlorophenoxy)propionic acid, 98%
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.