accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25148-68-9

IUPAC Name: dihydrogen N1-methylbenzene-1,2-diamine dichloride
Synonyms: dihydrogen N1-methylbenzene-1,2-diamine dichloride
Molecular Weight (g/mol): 195.09
Molecular Formula: C7H12Cl2N2
InChi Key: DKEONVNYXODZRQ-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CNC1=CC=CC=C1N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results